Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 010894.14
also known as 010894-14
Price (USD)/ Each
238.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
238.00
Add to cart
Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number010894.14
Price (USD)/ Each
238.00
-
Add to cart
Chemical Identifiers
CAS1370709-86-6
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.