Titanium(IV) oxide, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Titanium(IV) oxide, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium(IV) oxide, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 010897.18
also known as 010897-18
Price (USD)/ Each
675.65
Online exclusive
751.00 
Save 75.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
675.65
Online exclusive
751.00 
Save 75.35 (10%)
Add to cart
Titanium(IV) oxide, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number010897.18
Price (USD)/ Each
675.65
Online exclusive
751.00 
Save 75.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13463-67-7
IUPAC Namedioxotitanium
Molecular FormulaO2Ti
InChI KeyGWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILESO=[Ti]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Elemental AnalysisTitanium Ti: ≥58.95% (Theory 59.934%)
Total Metal Impurities≤0.0055% (≤55ppm)
FormPowder
Titanium(IV) oxide is used in paints, varnishes, paper, plastics printing inks, sun screen, fibers, rubber, cosmetic products, anti-fogging coatings, foodstuffs, pure titanium, glass, glass ceramics, memristor, electrical ceramics, catalysts, electric conductors and chemical intermediates. It exhibit excellent photocatalytic properties (Honda-Fujishima effect) In addition, titanium dioxide is also used to cleave protein at the site where proline is present.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only