Thermo Scientific Chemicals

Titanium(IV) oxide, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010897.30
250 g, Each
Thermo Scientific Chemicals

Titanium(IV) oxide, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010897.30
250 g, Each
Quantity
Catalog number: 010897.30
also known as 010897-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13463-67-7
IUPAC Name
dioxotitanium
Molecular Formula
O2Ti
InChI Key
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILES
O=[Ti]=O
Appearance (Color)
White
Elemental Analysis
Titanium Ti: ≥58.95% (Theory 59.934%)
Total Metal Impurities
≤0.0055% (≤55ppm)
Form
Powder

Description

Titanium(IV) oxide is used in paints, varnishes, paper, plastics printing inks, sun screen, fibers, rubber, cosmetic products, anti-fogging coatings, foodstuffs, pure titanium, glass, glass ceramics, memristor, electrical ceramics, catalysts, electric conductors and chemical intermediates. It exhibit excellent photocatalytic properties (Honda-Fujishima effect) In addition, titanium dioxide is also used to cleave protein at the site where proline is present.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text