Thermo Scientific Chemicals

Vanadium(V) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010904.04
2 g, Each
Thermo Scientific Chemicals

Vanadium(V) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010904.04
2 g, Each
Quantity
Catalog number: 010904.04
also known as 010904-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-62-1
IUPAC Name
[(dioxovanadio)oxy]dioxovanadium
Molecular Formula
O5V2
InChI Key
GNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILES
O=[V](=O)O[V](=O)=O
Total Metal Impurities
0.01% max.

Description

Vanadium pentoxide is the precursor to alloys of vanadium. It is widely used as an industrial catalyst for the manufacture of sulfuric acid, maleic anhydride, phthalic anhydride, adipic acid, acrylic acid, oxalic acid and anthraquinone. It finds application as a detector material in bolometers and microbolometer arrays for thermal imaging, and as an ethanol sensor in ppm (parts per million) levels. It is also used in vanadium redox batteries, which are used for energy storage in power facilities, such as wind farms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text