Thermo Scientific Chemicals

Holmium(III) oxide, REacton™, 99.995% (REO), Thermo Scientific Chemicals

Catalog number: 010910.14
25 g, Each
Thermo Scientific Chemicals

Holmium(III) oxide, REacton™, 99.995% (REO), Thermo Scientific Chemicals

Catalog number: 010910.14
25 g, Each
Quantity
Catalog number: 010910.14
also known as 010910-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12055-62-8
IUPAC Name
diholmium(3+) trioxidandiide
Molecular Formula
Ho2O3
InChI Key
OWCYYNSBGXMRQN-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Ho+3].[Ho+3]
Total Rare Earth Oxide Impurities
0.005% max.

Description

Holmium(III) oxide iacts as colorants which is used for cubic zirconia and glass, as a calibration standard for optical spectrophotometers, as a specialty catalyst, phosphor and a laser material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text