Thermo Scientific Chemicals

Lutetium(III) oxide, REacton™, 99.995% (metals basis), REM 50ppm, Thermo Scientific Chemicals

Catalog number: 010912.06
5 g, Each
Thermo Scientific Chemicals

Lutetium(III) oxide, REacton™, 99.995% (metals basis), REM 50ppm, Thermo Scientific Chemicals

Catalog number: 010912.06
5 g, Each
Quantity
Catalog number: 010912.06
also known as 010912-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12032-20-1
IUPAC Name
dilutetium(3+) trioxidandiide
Molecular Formula
Lu2O3
InChI Key
UGBIHFMRUDAMBY-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Lu+3].[Lu+3]
Comment
REM 50ppm
Total Metal Impurities
0.005% max.

Description

The product of Lutetium(III) oxide and palladium(II) oxide has a potential use in superconducting materials. It is an important raw material for laser crystals. It has specialized uses in ceramics, glass, phosphors, and lasers. Also used as a catalyst in cracking, alkylation, hydrogenation, and polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text