Scandium(III) oxide, REacton™, 99.998% (metals basis), REM 50ppm max, Thermo Scientific Chemicals
Scandium(III) oxide, REacton™, 99.998% (metals basis), REM 50ppm max, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Scandium(III) oxide, REacton™, 99.998% (metals basis), REM 50ppm max, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 010916.03
also known as 010916-03
Price (USD)/ Each
110.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
110.00
Add to cart
Scandium(III) oxide, REacton™, 99.998% (metals basis), REM 50ppm max, Thermo Scientific Chemicals
Catalog number010916.03
Price (USD)/ Each
110.00
-
Add to cart
Chemical Identifiers
CAS12060-08-1
IUPAC Namediscandium(3+) trioxidandiide
Molecular FormulaO3Sc2
InChI KeyHJGMWXTVGKLUAQ-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Sc+3].[Sc+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max., REM 50ppm max
A high melting white solid used in high-temperature systems (for its resistance to heat and thermal shock), electronic ceramics, and glass composition. Suitable for vacuum deposition applications. Also used to make scandium fluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only