Ytterbium(III) oxide, REacton™, 99.998% (REO), Thermo Scientific Chemicals
Ytterbium(III) oxide, REacton™, 99.998% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ytterbium(III) oxide, REacton™, 99.998% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 010919.06
also known as 010919-06
Price (USD)/ Each
80.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
80.00
Add to cart
Ytterbium(III) oxide, REacton™, 99.998% (REO), Thermo Scientific Chemicals
Catalog number010919.06
Price (USD)/ Each
80.00
-
Add to cart
Chemical Identifiers
CAS1314-37-0
IUPAC Namediytterbium(3+) trioxidandiide
Molecular FormulaO3Yb2
InChI KeyUZLYXNNZYFBAQO-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Yb+3].[Yb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.002% max.
It serves as a dopant for silicate mineral crystals in lasers, and a colorant for glasses and enamels. It is employed in the preparation of optical fibers, and special types of alloys. It is used in the preparation of chiral ytterbium complexes which are used as catalysts for enantioselective organic synthesis with high yields. It finds application in optic, electronic and ceramic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only