Thermo Scientific Chemicals

Ytterbium(III) oxide, REacton™, 99.998% (REO), Thermo Scientific Chemicals

Catalog number: 010919.14
25 g, Each
Thermo Scientific Chemicals

Ytterbium(III) oxide, REacton™, 99.998% (REO), Thermo Scientific Chemicals

Catalog number: 010919.14
25 g, Each
Quantity
Catalog number: 010919.14
also known as 010919-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-37-0
IUPAC Name
diytterbium(3+) trioxidandiide
Molecular Formula
O3Yb2
InChI Key
UZLYXNNZYFBAQO-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Yb+3].[Yb+3]
Total Rare Earth Oxide Impurities
0.002% max.

Description

It serves as a dopant for silicate mineral crystals in lasers, and a colorant for glasses and enamels. It is employed in the preparation of optical fibers, and special types of alloys. It is used in the preparation of chiral ytterbium complexes which are used as catalysts for enantioselective organic synthesis with high yields. It finds application in optic, electronic and ceramic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text