Thermo Scientific Chemicals

Calcium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 010923.A1
1 kg, Each
Thermo Scientific Chemicals

Calcium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 010923.A1
1 kg, Each
Quantity
Catalog number: 010923.A1
also known as 010923-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1305-78-8
IUPAC Name
calcium oxidandiide
Molecular Formula
CaO
InChI Key
BRPQOXSCLDDYGP-UHFFFAOYSA-N
SMILES
[O--].[Ca++]
Comment
May contain traces of Ca(OH)2 which disintegrates upon heating up to 120?
Total Metal Impurities
0.05% max.
Particle Size
-400 Mesh
Form
Powder

Description

It is used in cement, glass, steel, paper, chemical and petroleum industries. It acts as a key ingredient in the process of cement making. It is widely used industrially as a dehydrating agent and an absorbent. Further it finds use as a water softener and as a pH regulator for wastewater. It is useful to detect traces of water in hydrocarbons, and in flue-gas desulfurization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text