Nickel wire, 2.0mm (0.08in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Nickel wire, 2.0mm (0.08in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel wire, 2.0mm (0.08in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Form: Approx.28.0g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
5 cm
100 cm
Catalog number 010928.BU
also known as 010928-BU
Price (USD)/ Each
188.00
-
Add to cart
Quantity:
25 cm
Request bulk or custom format
Price (USD)/ Each
188.00
Add to cart
Nickel wire, 2.0mm (0.08in) dia, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number010928.BU
Price (USD)/ Each
188.00
-
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormWire, 2.0mm (0.08in) diameter.
Assay from Supplier's CofA≥99.995% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only