Iron wire, 0.1mm (0.004in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Iron wire, 0.1mm (0.004in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron wire, 0.1mm (0.004in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Form: approx. 0.06g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 m
10 m
Catalog number 010938.H2
also known as 010938-H2
Price (USD)/ Each
702.65
Online exclusive
781.00 
Save 78.35 (10%)
-
Add to cart
Quantity:
50 m
Request bulk or custom format
Price (USD)/ Each
702.65
Online exclusive
781.00 
Save 78.35 (10%)
Add to cart
Iron wire, 0.1mm (0.004in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number010938.H2
Price (USD)/ Each
702.65
Online exclusive
781.00 
Save 78.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialIron
ColorGray
CAS7439-89-6
Health Hazard 1H500
AppearanceGray wire
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only