Cobalt wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Cobalt wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 010946.BU
also known as 010946-BU
Price (USD)/ Each
152.65
Online exclusive
169.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
25 cm
Request bulk or custom format
Price (USD)/ Each
152.65
Online exclusive
169.00 
Save 16.35 (10%)
Add to cart
Cobalt wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number010946.BU
Price (USD)/ Each
152.65
Online exclusive
169.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.005%
FormWire, 1.0 mm (0.04 in.) dia
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only