Copper discs, 9.0^+0.127mm (0.354^+0.005in) dia x 0.8^+0.051mm (0.03^+0.002in) thick, 99.9995% (metals basis), Thermo Scientific Chemicals
Catalog number010951.09
Price (USD)/ Each
176.00
-
Add to cart
Specifications
Chemical Name or MaterialCopper discs
Name Note9.0±0.127mm (0.354±0.005 in.) dia. x 0.8±0.051mm (0.03±0.002 in.) thick
CAS7440-50-8
Recommended StorageAmbient temperatures
Physical FormDisc
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.