Copper shot, 6mm (0.24in) & down, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Copper shot, 6mm (0.24in) & down, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper shot, 6mm (0.24in) & down, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 010955.36
also known as 010955-36
Price (USD)/ Each
1,601.65
Online exclusive
1779.00 
Save 177.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
1,601.65
Online exclusive
1779.00 
Save 177.35 (10%)
Add to cart
Copper shot, 6mm (0.24in) & down, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Catalog number010955.36
Price (USD)/ Each
1,601.65
Online exclusive
1779.00 
Save 177.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only