Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 cm
10 cm
50 cm
Catalog number 010958.CB
also known as 010958-CB
Price (USD)/ Each
833.65
Online exclusive
926.00 
Save 92.35 (10%)
-
Add to cart
Quantity:
250 cm
Request bulk or custom format
Price (USD)/ Each
833.65
Online exclusive
926.00 
Save 92.35 (10%)
Add to cart
Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
Catalog number010958.CB
Price (USD)/ Each
833.65
Online exclusive
926.00 
Save 92.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment0.25mm (0.01in) dia
Total Metal Impurities0.003% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only