Copper wire, 0.1mm (0.004in) dia, hard, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Copper wire, 0.1mm (0.004in) dia, hard, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper wire, 0.1mm (0.004in) dia, hard, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals

Form: approx. 0.07g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 m
10 m
Catalog number 010971.H2
also known as 010971-H2
Price (USD)/ Each
400.65
Online exclusive
445.00 
Save 44.35 (10%)
-
Add to cart
Quantity:
50 m
Request bulk or custom format
Price (USD)/ Each
400.65
Online exclusive
445.00 
Save 44.35 (10%)
Add to cart
Copper wire, 0.1mm (0.004in) dia, hard, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Catalog number010971.H2
Price (USD)/ Each
400.65
Online exclusive
445.00 
Save 44.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0015% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only