Potassium hydroxide, 30% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 30% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hydroxide, 30% w/v aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number 010979.AP
also known as 010979-AP
Price (USD)/ Each
37.65
Online exclusive
42.20 
Save 4.55 (11%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
42.20 
Save 4.55 (11%)
Add to cart
Potassium hydroxide, 30% w/v aq. soln., Thermo Scientific Chemicals
Catalog number010979.AP
Price (USD)/ Each
37.65
Online exclusive
42.20 
Save 4.55 (11%)
-
Add to cart
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration (w/v)30% (w/v) aq. soln.
Potassium hydroxide acts as the precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only