Thermo Scientific Chemicals

Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010980.18
50 g, Each
Thermo Scientific Chemicals

Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010980.18
50 g, Each
Quantity
Catalog number: 010980.18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-23-3
IUPAC Name
potassium fluoride
Molecular Formula
FK
InChI Key
NROKBHXJSPEDAR-UHFFFAOYSA-M
SMILES
[F-].[K+]
Assay (special)
?2.5 % of Theoretical value (K)
Total Metal Impurities
≤0.01% (100 ppm)
Appearance (Color)
White
Form
Crystals

Description

Potassium fluoride is used in organic chemistry as a fluorinating agent to replace other halide ions like chorine, bromine etc. It is used as a catalyst in the manufacture of polyurethanes. Further, it is widely used for etching of glass, in disinfectant formulations and manufacture of optical glass. It is used in metal finishing, batteries, coatings and photographic chemicals. It is utilized for the study of ion-specific swelling and de-swelling of ampholytic polymer gels as well as in the measurement of electronic polarizabilities of ions in polymers of alkali halides. It finds application in the electronic industry as a metal surface treatment product. It is used as a preservative, a food additive, a catalyzer and a water absorbing agent. In the Finkelstein reaction, it is actively involved in the conversion of chlorocarbons to fluorocarbons using polar solvents like dimethyl formamide and ethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text