Thermo Scientific Chemicals

Perchloric acid, 70%, 99.9985% (metals basis), Thermo Scientific Chemicals

Catalog number: 010983.AK
250 mL, Each
Thermo Scientific Chemicals

Perchloric acid, 70%, 99.9985% (metals basis), Thermo Scientific Chemicals

Catalog number: 010983.AK
250 mL, Each
Quantity
Catalog number: 010983.AK
also known as 010983-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7601-90-3
IUPAC Name
perchloric acid
Molecular Formula
ClHO4
InChI Key
VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES
O[Cl](=O)(=O)=O
Total Metal Impurities
≤0.0015%
Concentration (weight/weight)
≥65%

Description

Perchloric acid is used as a precursor for ammonium perchlorate, which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis. It finds application in the computer industry for etching liquid crystal displays in critical electronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text