Thermo Scientific Chemicals

Lithium nitrate, anhydrous, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 010985.30
250 g, Each
Thermo Scientific Chemicals

Lithium nitrate, anhydrous, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 010985.30
250 g, Each
Quantity
Catalog number: 010985.30
also known as 010985-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-69-4
IUPAC Name
lithium(1+) nitrate
Molecular Formula
LiNO3
InChI Key
IIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES
[Li+].[O-][N+]([O-])=O
Total Metal Impurities
0.02% max.

Description

Lithium nitrate, anhydrous is used in the manufacture of red-colored fireworks and flares. has been proposed as a medium to store heat collected from the sun for cooking. A Fresnel lens would be used to melt solid lithium nitrate, which would then function as a 'solar battery', allowing heat to be redistributed later by convection

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text