Hydrofluoric acid, 99.99% (metals basis) 40% min, Thermo Scientific Chemicals
Hydrofluoric acid, 99.99% (metals basis) 40% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrofluoric acid, 99.99% (metals basis) 40% min, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010989.AC
Price (USD)/ Each
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS7664-39-3
IUPAC Namehydrogen fluoride
Molecular FormulaFH
InChI KeyKRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILESF
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)HF 40% min
Total Metal Impurities0.01% max.
Hydrofluoric acid is used in the glass etching, electronic and chemical industries. Hydrogen fluoride is mainly used in the production of organofluorine compounds, aluminum and chlorofluorocarbons. Hydrogen fluoride is also used for separating uranium isotopes, as a catalyst in the petroleum industry, and in stainless steel pickling. Hydrofluoric acid is used as a pickling agent and used in the semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only