Thermo Scientific Chemicals

Hydrochloric acid, 99.999999% (metals basis), 33% min, Thermo Scientific Chemicals

Catalog number: 010990.AC
25 mL, Each
Thermo Scientific Chemicals

Hydrochloric acid, 99.999999% (metals basis), 33% min, Thermo Scientific Chemicals

Catalog number: 010990.AC
25 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 010990.AC
also known as 010990-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-01-0
IUPAC Name
hydrogen chloride
Molecular Formula
ClH
InChI Key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES
Cl
Appearance (Color)
Colorless
Form
Liquid
Assay from Supplier's CofA
≥33.0%
Total Metal Impurities
≤0.000001%

Description

Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing. It finds use in the manufacture of polyvinyl chloride (PVC). Further, it is employed for regeneration of ion exchangers and in the tanning industry for the processing of leathers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text