Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Thermo Scientific Chemicals
Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Thermo Scientific Chemicals

Air Sensitive
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
500 mL
Catalog number 010991.AE
also known as 010991-AE
Price (USD)/ Each
182.65
Online exclusive
203.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
182.65
Online exclusive
203.00 
Save 20.35 (10%)
Add to cart
Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Thermo Scientific Chemicals
Catalog number010991.AE
Price (USD)/ Each
182.65
Online exclusive
203.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10035-10-6
IUPAC Namehydrogen bromide
Molecular FormulaBrH
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-N
SMILESBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Total Metal Impurities≤ 0.0001%
FormLiquid
Concentration (weight/weight)44-49%
Hydrobromic acid is used for the production of inorganic bromides of zinc, calcium, and sodium. It is used as a catalyst in some alkylation reactions. In the field of metallurgy, it is employed in the extraction of certain ores. Further, it is used as a reagent for the cleavage of certain ether linkages or as a brominating agent in organic transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only