Thermo Scientific Chemicals

Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 010991.AP
500 mL, Each
Thermo Scientific Chemicals

Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 010991.AP
500 mL, Each
Quantity
Catalog number: 010991.AP
also known as 010991-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10035-10-6
IUPAC Name
hydrogen bromide
Molecular Formula
BrH
InChI Key
CPELXLSAUQHCOX-UHFFFAOYSA-N
SMILES
Br
Appearance (Color)
Colorless to yellow
Total Metal Impurities
≤ 0.0001%
Concentration (weight/weight)
44-49%
Form
Liquid

Description

Hydrobromic acid is used for the production of inorganic bromides of zinc, calcium, and sodium. It is used as a catalyst in some alkylation reactions. In the field of metallurgy, it is employed in the extraction of certain ores. Further, it is used as a reagent for the cleavage of certain ether linkages or as a brominating agent in organic transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text