Thermo Scientific Chemicals

Cesium iodide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010992.22
100 g, Each
Thermo Scientific Chemicals

Cesium iodide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010992.22
100 g, Each
Quantity
Catalog number: 010992.22
also known as 010992-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-17-5
IUPAC Name
caesium(1+) iodide
Molecular Formula
CsI
InChI Key
XQPRBTXUXXVTKB-UHFFFAOYSA-M
SMILES
[I-].[Cs+]
Total Metal Impurities
0.001% max.

Description

Cesium iodide is used as an input phosphor in x-ray image intensifier tube of fluoroscopy equipment. It is also used as an equipment for electromagnetic calorimetry in experimental particle physics. It is often employed as a beamsplitter in Fourier Transform Infrared (FT-IR) spectrometers. Cesium iodide is an inorganic scintillator which is mainly used in experimental nuclear physics, high energy physics, nuclear medicine, nuclear tomography, environmental studies and many other fields of use. It is utilized in calorimeters and detectors and also employed as a phosphor in fluoroscopy equipments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text