Barium chloride, anhydrous, 99.998% (metals basis), Thermo Scientific Chemicals
Barium chloride, anhydrous, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium chloride, anhydrous, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
5 g
25 g
100 g
Catalog number 010995.02
also known as 010995-02
Price (USD)/ Each
25.65
Online exclusive
30.30 
Save 4.65 (15%)
-
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (USD)/ Each
25.65
Online exclusive
30.30 
Save 4.65 (15%)
Add to cart
Barium chloride, anhydrous, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number010995.02
Price (USD)/ Each
25.65
Online exclusive
30.30 
Save 4.65 (15%)
-
Add to cart
Chemical Identifiers
CAS10361-37-2
IUPAC Namebarium(2+) dichloride
Molecular FormulaBaCl2
InChI KeyWDIHJSXYQDMJHN-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder, may contain small lumps
Total Metal Impurities≤0.002% (20 ppm)
Barium chloride is used in manufacture of pigments, fire works, other barium salts, fireworks , hardening of steel, heat treatment salts, purification of brine solution in caustic chlorine plants, lubrication oil additive, textile dye, pigments, white leather, aluminum refining, boiling water treatment and porcelain enamels for sheet steel. It is also used to remove sulfate ion in some electrolytic plants, caustic soda, magnesium metal or sodium metal. It is also used as a component in a flux used to prevent oxidation of molten magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only