Thermo Scientific Chemicals

Barium chloride, anhydrous, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010995.14
25 g, Each
Thermo Scientific Chemicals

Barium chloride, anhydrous, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010995.14
25 g, Each
Quantity
Catalog number: 010995.14
also known as 010995-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10361-37-2
IUPAC Name
barium(2+) dichloride
Molecular Formula
BaCl2
InChI Key
WDIHJSXYQDMJHN-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Ba++]
Appearance (Color)
White
Form
Powder, may contain small lumps
Total Metal Impurities
≤0.002% (20 ppm)

Description

Barium chloride is used in manufacture of pigments, fire works, other barium salts, fireworks , hardening of steel, heat treatment salts, purification of brine solution in caustic chlorine plants, lubrication oil additive, textile dye, pigments, white leather, aluminum refining, boiling water treatment and porcelain enamels for sheet steel. It is also used to remove sulfate ion in some electrolytic plants, caustic soda, magnesium metal or sodium metal. It is also used as a component in a flux used to prevent oxidation of molten magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text