Zinc selenide, 99.999% (metals basis), Thermo Scientific Chemicals
Zinc selenide, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc selenide, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 010999.18
also known as 010999-18
Price (USD)/ Each
293.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
293.00
Add to cart
Zinc selenide, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number010999.18
Price (USD)/ Each
293.00
-
Add to cart
Chemical Identifiers
CAS1315-09-9
IUPAC Namezinc(2+) selandiide
Molecular FormulaSeZn
InChI KeyGTLQJUQHDTWYJC-UHFFFAOYSA-N
SMILES[Zn++].[Se--]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Zinc selenide (ZnSe) is used as an infrared optical material with wide transmission wavelength range. It is also used as an entrance optic in the new range of in-ear clinical thermometers. ZnSe is used as a semiconductor material for thin film devices, and n-type windows layer for thin film heterojunction solar cells. ZnSe activated with tellurium is a scintillator suitable for matching with photodiodes. It is used in x-ray and gamma ray detectors. Zinc selenide films are useful in photovoltaic cells and solar conversion cells. The material can be doped with n-type and p-type doping. It is used to form light-emitting diodes and diode lasers. ZnSe doped with chromium finds use as an IR laser gain medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only