Thermo Scientific Chemicals

Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals

Catalog number: 011000.AB
10 mL, Each
Thermo Scientific Chemicals

Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals

Catalog number: 011000.AB
10 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 011000.AB
also known as 011000-AB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7664-93-9
IUPAC Name
sulfuric acid
Molecular Formula
H2O4S
InChI Key
QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O
Appearance (Color)
Clear colorless
Assay from Supplier's CofA
≥92.0%
Total Metal Impurities
≤0.0001%
Form
Liquid

Description

Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, insecticides, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text