Thermo Scientific Chemicals

Sodium acetate, anhydrous, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 011005.06
5 g, Each
Thermo Scientific Chemicals

Sodium acetate, anhydrous, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 011005.06
5 g, Each
Quantity
Catalog number: 011005.06
also known as 011005-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-09-3
IUPAC Name
sodium acetate
Molecular Formula
C2H3NaO2
InChI Key
VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES
[Na+].CC([O-])=O
Appearance (Color)
White
Form
Powder
Assay (special)
Sodium (Na): 25.5-30.5%
Total Metal Impurities
≤0.003% (30 ppm)

Description

Sodium acetate is used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, as a plating agent, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography. It is used as a base in several organic reactions in research laboratories and industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text