Orion™ Conductivity Standards and Solutions
Orion™ Conductivity Standards and Solutions
Actual product may vary
Orion™ Conductivity Standards and Solutions
Orion™ Conductivity Standards and Solutions
Thermo Scientific™

Orion™ Conductivity Standards and Solutions

Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackagingConductivity
011008Bottle100 μS/cm
011001Bottle
011007Bottle1413 μS/cm
011005Bottles111.9 mS/cm
011006Bottle12.9 mS/cm
990106Bottle12.9 mS
01100910Pouch147 μS/cm
01100510Pouch111.9 mS/cm
01100710Pouch1413 μS/cm
01100610Pouch12.9 mS/cm
Catalog number 011008
Price (HKD)
-
Request A Quote
Packaging:
Bottle
Conductivity:
100 μS/cm
Ask our AI about this Product

Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.

Conductivity Standards

  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
  • For use with Thermo Scientific™ Orion™ Conductivity Probes

Warranty: Out of Box

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Boiling Point100°C
ColorClear
Conductivity100 μS/cm
DescriptionConductivity Standard
For Use With (Equipment)Orion pH Meter
PackagingBottle
Quantity5 x 60 mL
SolubilitySoluble
Solution TypeStandard Solution
Traceability to NISTYes
TypeConductivity Standard
Concentration47 ppm as NaCl
pH4.75 to 7.75
Unit SizePack of 5
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
0004506382Certificate of AnalysisJun 12, 2025011006
DY3Certificate of AnalysisMar 03, 2025011008
DY2Certificate of AnalysisMar 03, 2025011008
DY2ACertificate of AnalysisMar 03, 2025011008
CQ1Certificate of AnalysisFeb 12, 2025011006
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection