Potassium nitrate, 99.994% (metals basis), Thermo Scientific Chemicals
Potassium nitrate, 99.994% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium nitrate, 99.994% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011008.22
also known as 011008-22
Price (USD)/ Each
211.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
249.00 
Save 37.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
211.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
249.00 
Save 37.35 (15%)
Add to cart
Potassium nitrate, 99.994% (metals basis), Thermo Scientific Chemicals
Catalog number011008.22
Price (USD)/ Each
211.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
249.00 
Save 37.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7757-79-1
IUPAC Namepotassium nitrate
Molecular FormulaKNO3
InChI KeyFGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES[K+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.006% max.
Potassium nitrate has several applications. It is used as an oxidizer, food preservative, fertilizer, and an active ingredient of condensed aerosol fire suppression systems. It is a powerful nitrating agent in sulfuric acid medium. In addition, it also used as an electrolyte in a salt bridge and thermal storage medium in power generation systems. Ternary salts containing potassium nitrate have been reported to improve the heat storage capacity in the molten salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only