Calcium fluoride, 99%, Thermo Scientific Chemicals
Calcium fluoride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium fluoride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 011022.09
also known as 011022-09
Price (USD)/ Each
57.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
57.00
Add to cart
Calcium fluoride, 99%, Thermo Scientific Chemicals
Catalog number011022.09
Price (USD)/ Each
57.00
-
Add to cart
Chemical Identifiers
CAS7789-75-5
IUPAC Namecalcium difluoride
Molecular FormulaCaF2
InChI KeyWUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES[F-].[F-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
It finds its application in spectroscopy, electronics, lasers, high-temperature dry-film lubricants, emery wheels, certain cements, phosphors, paint pigment, catalyst in wood preservatives, and optical equipment. Calcium fluoride is used as component of electrolyte, fluxing agent for aluminum metallurgy. It is used in glass and fluorescent lamps industry. It has dental applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only