Thermo Scientific Chemicals

Calcium fluoride, 99%, Thermo Scientific Chemicals

Catalog number: 011022.30
250 g, Each
Thermo Scientific Chemicals

Calcium fluoride, 99%, Thermo Scientific Chemicals

Catalog number: 011022.30
250 g, Each
Quantity
Catalog number: 011022.30
also known as 011022-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-75-5
IUPAC Name
calcium difluoride
Molecular Formula
CaF2
InChI Key
WUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES
[F-].[F-].[Ca++]
Assay (unspecified)
98.6% min

Description

It finds its application in spectroscopy, electronics, lasers, high-temperature dry-film lubricants, emery wheels, certain cements, phosphors, paint pigment, catalyst in wood preservatives, and optical equipment. Calcium fluoride is used as component of electrolyte, fluxing agent for aluminum metallurgy. It is used in glass and fluorescent lamps industry. It has dental applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text