Thermo Scientific™

Sodium fluoride, Optical Grade, Thermo Scientific Chemicals

Catalog number: 011023.18
50 g, Each
Thermo Scientific™

Sodium fluoride, Optical Grade, Thermo Scientific Chemicals

Catalog number: 011023.18
50 g, Each
Quantity
Catalog number: 011023.18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7681-49-4
IUPAC Name
sodium fluoride
Molecular Formula
FNa
InChI Key
PUZPDOWCWNUUKD-UHFFFAOYSA-M
SMILES
[F-].[Na+]
Total Metal Impurities
0.1% max.

Description

It has medical and other applications including complexing of magnesium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text