Thermo Scientific Chemicals

Rhodium(III) chloride hydrate, Rh 38.0-45.5%, Thermo Scientific Chemicals

Catalog number: 011032.06
5 g, Each
Thermo Scientific Chemicals

Rhodium(III) chloride hydrate, Rh 38.0-45.5%, Thermo Scientific Chemicals

Catalog number: 011032.06
5 g, Each
Quantity
Catalog number: 011032.06
also known as 011032-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20765-98-4
IUPAC Name
rhodium(3+) trichloride
Molecular Formula
Cl3Rh
InChI Key
SONJTKJMTWTJCT-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Rh+3]
Assay (unspecified)
Rh 38.0-45.5%

Description

Rhodium(III) chloride hydrate is used as a catalyst to prepare acetic acid. It is also used as a catalyst for reduction, oxidation of alkenes, hydration of acetylene and isomerization of alkenes. It is involved in the hydrosilylation of alfa,beta-unsaturated esters to form dimethylketene trimethylsilyl acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text