Thermo Scientific Chemicals

trans-Diamminedichloropalladium(II), Premion™, 99.95% (metals basis), Pd 49.9% min, Thermo Scientific Chemicals

Catalog number: 011037.02
0.5 g, Each
Thermo Scientific Chemicals

trans-Diamminedichloropalladium(II), Premion™, 99.95% (metals basis), Pd 49.9% min, Thermo Scientific Chemicals

Catalog number: 011037.02
0.5 g, Each
Quantity
Catalog number: 011037.02
also known as 011037-02
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14323-43-4
IUPAC Name
palladium(2+) diamine dichloride
Molecular Formula
Cl2H6N2Pd
InChI Key
WIEZTXFTOIBIOC-UHFFFAOYSA-L
SMILES
N.N.[Cl-].[Cl-].[Pd++]
Total Metal Impurities
0.05% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text