Thermo Scientific Chemicals

Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min, Thermo Scientific Chemicals

Catalog number: 011046.06
5 g, Each
Thermo Scientific Chemicals

Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min, Thermo Scientific Chemicals

Catalog number: 011046.06
5 g, Each
Quantity
Catalog number: 011046.06
also known as 011046-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13820-41-2
IUPAC Name
platinum(2+) diammonium tetrachloride
Molecular Formula
Cl4H8N2Pt
InChI Key
QJIMNDWDOXTTBR-UHFFFAOYSA-L
SMILES
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt++]
Total Metal Impurities
0.1% max., Pt 51% min

Description

Ammonium tetrachloroplatinate(II) is used in spectral analysis standard. It is also used in the preparation of platinum sponge and platinum catalytic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text