Thermo Scientific Chemicals

Sodium hexachloroplatinate(IV) hexahydrate, Thermo Scientific Chemicals

Catalog number: 011047.06
5 g, Each
Thermo Scientific Chemicals

Sodium hexachloroplatinate(IV) hexahydrate, Thermo Scientific Chemicals

Catalog number: 011047.06
5 g, Each
Quantity
Catalog number: 011047.06
also known as 011047-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19583-77-8
IUPAC Name
disodium hexachloroplatinum hexahydrate
Molecular Formula
Cl6H12Na2O6Pt
InChI Key
XGFSWVNCHVSCSP-UHFFFAOYSA-H
SMILES
O.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl
Appearance (Color)
Orange
Assay from Supplier's CofA
31.30-37.00% w/w Platinum
Form
Crystals

Description

Catalyst.Sodium hexachloroplatinate(IV) hexahydrate is used as a catalyst. It acts as a source of platinum in the preparation of platinum complex. It is also used in the synthesis of platinum nanoparticles using glutathione as a reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text