Potassium tetrachloroplatinate(II), 99.9% (metals basis), Pt 46.0% min, Thermo Scientific Chemicals
Potassium tetrachloroplatinate(II), 99.9% (metals basis), Pt 46.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium tetrachloroplatinate(II), 99.9% (metals basis), Pt 46.0% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 011048.03
also known as 011048-03
Price (USD)/ Each
142.65
Online exclusive
158.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
142.65
Online exclusive
158.00 
Save 15.35 (10%)
Add to cart
Potassium tetrachloroplatinate(II), 99.9% (metals basis), Pt 46.0% min, Thermo Scientific Chemicals
Catalog number011048.03
Price (USD)/ Each
142.65
Online exclusive
158.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10025-99-7
IUPAC Namedipotassium tetrachloroplatinumdiuide
Molecular FormulaCl4H4K2Pt
InChI KeyJCAXKWJYKUEOFO-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[K+].[K+].[Pt--]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
FormCrystals or crystalline powder
Assay (special)Pt: ≥46.0%
Total Metal Impurities≤0.1%
Potassium tetrachloroplatinate solution is used in the preparation of platinum (Pt) nanoparticles. Ligands like ammonia or triphenyl phosphine can able to replace the chloride ligands to afford various derivatives. The replacement by ammonia results in the preparation of cisplatin, which is used in the cancer treatment. Further, it is used as catalyst in the hydroarylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only