Thermo Scientific Chemicals

Platinum(IV) oxide hydrate, 99.9% (metals basis), Pt 71-75%, Thermo Scientific Chemicals

Catalog number: 011049.06
5 g, Each
Thermo Scientific Chemicals

Platinum(IV) oxide hydrate, 99.9% (metals basis), Pt 71-75%, Thermo Scientific Chemicals

Catalog number: 011049.06
5 g, Each
Quantity
Catalog number: 011049.06
also known as 011049-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52785-06-5
IUPAC Name
dioxoplatinum
Molecular Formula
O2Pt
InChI Key
YKIOKAURTKXMSB-UHFFFAOYSA-N
SMILES
O=[Pt]=O
Total Metal Impurities
0.1% max.

Description

Platinum(IV) oxide hydrate is used in the conversion of ketones to methylenes by hydrogenolysis of enol triflates under neutral conditions. It is used as a catalyst for hydrosilylation of olefins with various alkyl dialkoxy silanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text