Dihydrogen hexahydroxyplatinate(IV), 99.9% (metals basis), Pt 61.0% min, Thermo Scientific Chemicals
Dihydrogen hexahydroxyplatinate(IV), 99.9% (metals basis), Pt 61.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dihydrogen hexahydroxyplatinate(IV), 99.9% (metals basis), Pt 61.0% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
1 g
5 g
Catalog number 011052.02
also known as 011052-02
Price (USD)/ Each
98.00
-
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (USD)/ Each
98.00
Add to cart
Dihydrogen hexahydroxyplatinate(IV), 99.9% (metals basis), Pt 61.0% min, Thermo Scientific Chemicals
Catalog number011052.02
Price (USD)/ Each
98.00
-
Add to cart
Chemical Identifiers
CAS51850-20-5
IUPAC Nameplatinum(4+) dihydrogen hexahydroxide
Molecular FormulaH8O6Pt
InChI KeyILXDKMTYQJUXAB-UHFFFAOYSA-J
SMILES[H+].[H+].[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Pt+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pt 61.0% min
Total Metal Impurities0.1% max.
Dihydrogen hexahydroxyplatinate(IV) is used as a precursor to produce platinum dioxide (II), a catalyst of hydrogenation and hydrogenolysis of carbonyl group or C-Cl linkages in acyl chlorides. It also acts as a precursor to produce other platinum compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only