Thermo Scientific Chemicals

Calcium fluoride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 011055.30
250 g, Each
Thermo Scientific Chemicals

Calcium fluoride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 011055.30
250 g, Each
Quantity
Catalog number: 011055.30
also known as 011055-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-75-5
IUPAC Name
calcium difluoride
Molecular Formula
CaF2
InChI Key
WUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES
[F-].[F-].[Ca++]
Total Metal Impurities
≤0.5% (5000 ppm)
Form
Powder
Appearance (Color)
White

Description

Calcium fluoride is the commercially important source of fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers. It is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text