Copper powder, spherical, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Copper powder, spherical, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper powder, spherical, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 x 1 kg
Catalog number 011070.30
also known as 011070-30
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.20 
Save 6.55 (14%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.20 
Save 6.55 (14%)
Add to cart
Copper powder, spherical, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number011070.30
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.20 
Save 6.55 (14%)
-
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder, -100 mesh
Total Metal Impurities≤0.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only