Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number 011090.30
Price (USD)/ Each
377.65
Online exclusive
419.00 
Save 41.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
377.65
Online exclusive
419.00 
Save 41.35 (10%)
Add to cart
Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number011090.30
Price (USD)/ Each
377.65
Online exclusive
419.00 
Save 41.35 (10%)
-
Add to cart
Chemical Identifiers
CAS123334-20-3
IUPAC Nameoxovanadiumbis(ylium) sulfate
Molecular FormulaO5SV
InChI KeyUUUGYDOQQLOJQA-UHFFFAOYSA-L
SMILESO=[V++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Vanadium(IV) sulfate oxide hydrate is used as a catalyst for the oxidative aromatization of cyclohexenone. It is also used as a component in some food supplements. It is one of the intermediate during the extraction of vanadium from petroleum residues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only