Vanadium(V) oxide, 99.6% min, Thermo Scientific Chemicals
Vanadium(V) oxide, 99.6% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium(V) oxide, 99.6% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Catalog number 011094.09
also known as 011094-09
Price (USD)/ Each
32.65
Online exclusive
35.90 
Save 3.25 (9%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
32.65
Online exclusive
35.90 
Save 3.25 (9%)
Add to cart
Vanadium(V) oxide, 99.6% min, Thermo Scientific Chemicals
Catalog number011094.09
Price (USD)/ Each
32.65
Online exclusive
35.90 
Save 3.25 (9%)
-
Add to cart
Chemical Identifiers
CAS1314-62-1
IUPAC Name[(dioxovanadio)oxy]dioxovanadium
Molecular FormulaO5V2
InChI KeyGNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILESO=[V](=O)O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow-orange
FormGranules
Assay from Supplier's CofA≥99.6% (V₂O₅)
Vanadium pentoxide is the precursor to alloys of vanadium. It is widely used as an industrial catalyst for the manufacture of sulfuric acid, maleic anhydride, phthalic anhydride, adipic acid, acrylic acid, oxalic acid and anthraquinone. It finds application as a detector material in bolometers and microbolometer arrays for thermal imaging, and as an ethanol sensor in ppm (parts per million) levels. It is also used in vanadium redox batteries, which are used for energy storage in power facilities, such as wind farms. Additionally, it is used as ceramics colorant, as dye and also as a detector material in bolometers in the field of thermal imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only