Vanadium foil, 1.0mm (0.04in) thick, 99.5% (metals basis), Thermo Scientific Chemicals
Vanadium foil, 1.0mm (0.04in) thick, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium foil, 1.0mm (0.04in) thick, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
50 x 200 mm
Catalog number 011097.FI
also known as 011097-FI
Price (USD)/ Each
288.00
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
288.00
Add to cart
Vanadium foil, 1.0mm (0.04in) thick, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number011097.FI
Price (USD)/ Each
288.00
-
Add to cart
Chemical Identifiers
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Metallic grey
FormFoil, 1.0 mm thick
Total Metal Impurities≤0.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only