Rubidium carbonate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals
Rubidium carbonate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rubidium carbonate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number 011105.14
also known as 011105-14
Price (USD)/ Each
414.65
Online exclusive
460.00 
Save 45.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
414.65
Online exclusive
460.00 
Save 45.35 (10%)
Add to cart
Rubidium carbonate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals
Catalog number011105.14
Price (USD)/ Each
414.65
Online exclusive
460.00 
Save 45.35 (10%)
-
Add to cart
Chemical Identifiers
CAS584-09-8
IUPAC Namedirubidium(1+) carbonate
Molecular FormulaCO3Rb2
InChI KeyWPFGFHJALYCVMO-UHFFFAOYSA-L
SMILES[Rb+].[Rb+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.025% max.
It finds it application as a raw material for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only