Thermo Scientific Chemicals

Cesium sulfate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 011112.09
10 g, Each
Thermo Scientific Chemicals

Cesium sulfate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 011112.09
10 g, Each
Quantity
Catalog number: 011112.09
also known as 011112-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-54-9
IUPAC Name
dicaesium(1+) sulfate
Molecular Formula
Cs2O4S
InChI Key
FLJPGEWQYJVDPF-UHFFFAOYSA-L
SMILES
[Cs+].[Cs+].[O-]S([O-])(=O)=O
Total Metal Impurities
0.003% max.

Description

Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is also used for crystal growing purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text